ADV428293649 AChemBlock

cis-3-Amino-1-(trifluoromethyl)cyclobutan-1-ol hydrochloride

  • MDL number MFCD23106129
  • Cas Number : 1408075-93-3
  • Molecular Weight : 191.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/cis-3-amino-1-trifluoromethyl-cyclobutan-1-ol-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9022
pictograms GHS07
hstatements H302;H313;H319; H335
Availability details for
Quantity
  • Availability
  • Add to Cart