ADV428293653 AChemBlock

5-tert-Butyl-3-isothiazolamine

  • MDL number MFCD18451894
  • Cas Number : 127024-28-6
  • Molecular Weight : 156.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-tert-butyl-3-isothiazolamine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9026
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart