ADV428293657 AChemBlock

2,2-Dimethyl-3-oxocyclobutanecarboxylic acid

  • MDL number MFCD23106160
  • Cas Number : 3183-43-5
  • Molecular Weight : 142.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-2-dimethyl-3-oxocyclobutanecarboxylic-acid.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9030
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart