ADV428293664 AChemBlock

7-Boc-3-oxa-7,9-diazabicyclo[3.3.1]nonane

  • MDL number MFCD17016706
  • Cas Number : 864448-41-9
  • Molecular Weight : 228.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/7-boc-3-oxa-7-9-diazabicyclo-3-3-1-nonane.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9037
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart