ADV428293665 AChemBlock

cis-4,4-Difluorooctahydrocyclopenta[c]pyrrole

  • MDL number MFCD12198728
  • Cas Number : 1251008-46-4
  • Molecular Weight : 147.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/cis-4-4-difluorooctahydrocyclopenta-c-pyrrole.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9038
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart