ADV428293671 AChemBlock

(3R)-2-Azabicyclo[2.2.2]octane-3-carboxylic acid

  • MDL number MFCD23106232
  • Cas Number : 146144-65-2
  • Molecular Weight : 155.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/3r-2-azabicyclo-2-2-2-octane-3-carboxylic-acid.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9044
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart