ADV428293672 AChemBlock

3-Nitro-4,5,6,7-tetrahydropyrazolo[1,5-a]pyrimidine

  • MDL number MFCD23106242
  • Cas Number : 148777-84-8
  • Molecular Weight : 168.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-nitro-4-5-6-7-tetrahydropyrazolo-1-5-a-pyrimidine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9045
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart