ADV428293677 AChemBlock

1-Boc-(3aS,6aS)-octahydropyrrolo[3,4-b]pyrrole

  • Cas Number : 1018443-32-7
  • Molecular Weight : 212.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl (3aS,6aS)-hexahydropyrrolo[3,4-b]pyrrole-1(2H)-carboxylate
Inchi Code 1S/C11H20N2O2/c1-11(2,3)15-10(14)13-5-4-8-6-12-7-9(8)13/h8-9,12H,4-7H2,1-3H3/t8-,9+/m0/s1
Inchi Key OIKZNKIPPFFTKA-DTWKUNHWSA-N

Description

Synonym rac-tert-butyl (3ar,6ar)-hexahydropyrrolo[3,4-b]pyrrole-1(2h)-carboxylate
Purity 0.97
Shipping Temperature Room Temperature
Country of Origin CN
Product Link https://www.achemblock.com/1-boc-3as-6as-octahydropyrrolo-3-4-b-pyrrole.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/ac_sds?Catalog=I-9050
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart