ADV428293680 AChemBlock

(3R)-1-(Oxetan-3-yl)piperidin-3-amine

  • MDL number MFCD20233075
  • Cas Number : 1349700-06-6
  • Molecular Weight : 156.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/3r-1-oxetan-3-yl-piperidin-3-amine.html

Safety Information

signalword Warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9054
pictograms GHS07
hstatements H302;H313;H319; H335
Availability details for
Quantity
  • Availability
  • Add to Cart