ADV428293686 AChemBlock

3-Ethyl-3-oxetanecarbonitrile

  • MDL number MFCD22575230
  • Cas Number : 1414513-71-5
  • Molecular Weight : 111.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/3-ethyl-3-oxetanecarbonitrile.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9060
pictograms GHS02; GHS07
hstatements H226;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart