ADV428293688 AChemBlock

5-Bromo-4-chloro-6-methyl-2-(methylthio)pyrimidine

  • MDL number MFCD00834124
  • Cas Number : 17119-74-3
  • Molecular Weight : 253.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/5-bromo-4-chloro-6-methyl-2-methylthio-pyrimidine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9062
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart