ADV428293689 AChemBlock

cis-tert-Butyl N-2-hydroxycyclopentyl]carbamate

  • MDL number MFCD11977684
  • Cas Number : 155837-16-4
  • Molecular Weight : 201.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/cis-tert-butyl-n-2-hydroxycyclopentyl-carbamate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9063
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart