ADV428293697 AChemBlock

(2R)-1,4-Dioxane-2-methanamine hydrochloride

  • MDL number MFCD27918519
  • Cas Number : 1523541-84-5
  • Molecular Weight : 153.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2r-1-4-dioxane-2-methanamine-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9071
pictograms GHS07
hstatements H302;H313;H319; H335
Availability details for
Quantity
  • Availability
  • Add to Cart