ADV428293702 AChemBlock

2-(2,5-Difluorophenyl)pyrrolidine

  • MDL number MFCD05189458
  • Cas Number : 886503-15-7
  • Molecular Weight : 183.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-2-5-difluorophenyl-pyrrolidine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9076
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart