ADV428293705 AChemBlock

1-(Bromomethyl)-3-isopropylbenzene

  • MDL number MFCD15527192
  • Cas Number : 75369-42-5
  • Molecular Weight : 213.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/1-bromomethyl-3-isopropylbenzene.html

Safety Information

signalword Danger
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9079
pictograms GHS06; GHS07
hstatements H302;H314;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart