ADV428293709 AChemBlock

5-Bromo-1,3-dihydro-benzo(C)thiophene 2,2-dioxide

  • MDL number MFCD00666887
  • Cas Number : 351005-12-4
  • Molecular Weight : 247.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-bromo-1-3-dihydro-benzo-c-thiophene-2-2-dioxide.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9083
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart