ADV428293712 AChemBlock

2,4-Dichloro-5,6,7,8-tetrahydroquinoline

  • MDL number MFCD23135262
  • Cas Number : 858279-01-3
  • Molecular Weight : 202.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-4-dichloro-5-6-7-8-tetrahydroquinoline.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9086
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart