ADV428293717 AChemBlock

(2S)-Alpha,Alpha-dimethyl-2-pyrrolidinemethanol

  • MDL number MFCD11975452
  • Cas Number : 92053-25-3
  • Molecular Weight : 129.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2s-alpha-alpha-dimethyl-2-pyrrolidinemethanol.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9091
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart