ADV428293719 AChemBlock

2-Bromo-6,7-dihydrothieno[3,2-c]pyridin-4(5H)-one

  • MDL number MFCD11656583
  • Cas Number : 1078150-17-0
  • Molecular Weight : 232.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-bromo-6-7-dihydrothieno-3-2-c-pyridin-4-5h-one.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9093
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart