ADV428293720 AChemBlock

5-Bromo-3,3-difluoro-1,3-dihydro-1-methyl-2H-indol-2-one

  • MDL number MFCD27988080
  • Cas Number : 1630906-88-5
  • Molecular Weight : 262.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-bromo-3-3-difluoro-1-3-dihydro-1-methyl-2h-indol-2-one.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9094
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart