ADV465748107 AChemBlock

6-tert-Butyl 2-methyl 6-azaspiro[3.4]octane-2,6-dicarboxylate

  • MDL number MFCD28501606
  • Cas Number : 203662-61-7
  • Molecular Weight : 269.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/6-tert-butyl-2-methyl-6-azaspiro-3-4-octane-2-6-dicarboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9557
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart