ADV465748112 AChemBlock

(2S)-2-(Methoxymethyl)azetidine hemioxalate

  • MDL number MFCD27988020
  • Cas Number : 1523530-72-4
  • Molecular Weight : 292.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form Light yellow solid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/2s-2-methoxymethyl-azetidine-hemioxalate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6501
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart