ADV465748118 AChemBlock

1-Oxa-7-azaspiro[3.5]nonane hemioxalate

  • MDL number MFCD23702009
  • Cas Number : 1523617-84-6
  • Molecular Weight : 344.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1-oxa-7-azaspiro-3-5-nonane-hemioxalate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=A-971-1
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart