ADV465748119 AChemBlock

methyl (S)-3-cyclopropyl-3-(3-hydroxyphenyl)propanoate

  • MDL number MFCD27956898
  • Cas Number : 1142223-08-2
  • Molecular Weight : 220.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/benzenepropanoic-acid-beta-cyclopropyl-3-hydroxy-methyl-ester-betas.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7634
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart