ADV465748122 AChemBlock

8-(tert-butoxycarbonyl)-8-azabicyclo[3.2.1]octane-3-carboxylic acid

  • MDL number MFCD17214354
  • Cas Number : 1222996-05-5
  • Molecular Weight : 255.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/8-azabicyclo-3-2-1-octane-3-8-dicarboxylic-acid-8-1-1-dimethylethyl-ester-3-endo.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7837
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart