ADV465748123 AChemBlock

4-Methoxy-hexahydro-1H-azepine hydrochloride

  • MDL number MFCD21641983
  • Cas Number : 1408076-34-5
  • Molecular Weight : 165.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/4-methoxy-hexahydro-1h-azepine-hydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2753-1
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart