ADV465748154 AChemBlock

(2R,3R)-Benzyl 3-amino-2-methylpiperidine-1-carboxylate

  • MDL number MFCD22689535
  • Cas Number : 1044641-49-7
  • Molecular Weight : 248.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/2r-3r-benzyl-3-amino-2-methylpiperidine-1-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=H-8298
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart