ADV465748156 AChemBlock

(R)-3-Hydroxy-4-methoxy-4-oxobutanoic acid

  • MDL number MFCD16251425
  • Cas Number : 83540-94-7
  • Molecular Weight : 148.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8 °C
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/r-3-hydroxy-4-methoxy-4-oxobutanoic-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=H-8314
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart