ADV465748164 AChemBlock

4-Bromo-1H-pyrrolo[3,2-c]pyridine

  • MDL number MFCD08690131
  • Cas Number : 1000342-68-6
  • Molecular Weight : 197.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/4-bromo-1h-pyrrolo-3-2-c-pyridine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=H-8332
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart