ADV465748167 AChemBlock

(S)-2-Bromomethyl-pyrrolidine-1-carboxylic acid tert-butyl ester

  • MDL number MFCD10566043
  • Cas Number : 128542-75-6
  • Molecular Weight : 264.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/s-2-bromomethyl-pyrrolidine-1-carboxylic-acid-tert-butyl-ester.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=H-8346
pictograms GHS07
hstatements H303;H313;H333
Availability details for
Quantity
  • Availability
  • Add to Cart