ADV465748174 AChemBlock

1-(3-Bromopropyl)-4-(2,2,2- trifluoroethyl)piperazine

  • MDL number MFCD17262967
  • Cas Number : 1343109-33-0
  • Molecular Weight : 289.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/1-3-bromopropyl-4-2-2-2-trifluoroethyl-piperazine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=H-8360
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart