ADV465748180 AChemBlock

1-(3-Bromopropyl)-4-methylpiperazine hydrobromide

  • MDL number MFCD03426155
  • Cas Number : 88806-07-9
  • Molecular Weight : 302.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/1-3-bromopropyl-4-methylpiperazine-hydrobromide.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=H-8382
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart