ADV465748183 AChemBlock

6-Amino-3,4-dihydro-1,3-dimethyl-2(1H)-quinazolinone

  • MDL number MFCD22190706
  • Cas Number : 233775-53-6
  • Molecular Weight : 191.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/6-amino-3-4-dihydro-1-3-dimethyl-2-1h-quinazolinone.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=H-8390
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart