ADV465748185 AChemBlock

3-((3S,4S)-4-methyl-3-(methyl(7h-pyrrolo[2,3-d]pyrimidin-4-yl)amino)piperidin-1-yl)-3-oxopropanenitrile

  • MDL number MFCD11035920
  • Cas Number : 1092578-47-6
  • Molecular Weight : 312.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/3-3s-4s-4-methyl-3-methyl-7h-pyrrolo-2-3-d-pyrimidin-4-yl-amino-piperidin-1-yl-3-oxopropanenitrile.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=H-8396
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart