ADV465748194 AChemBlock

2-Bromo-4,6-difluoronitrobenzene

  • MDL number MFCD04112501
  • Cas Number : 884494-38-6
  • Molecular Weight : 237.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/2-bromo-4-6-difluoronitrobenzene.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=H-8407
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart