ADV465748213 AChemBlock

3-(1-Methylethoxy)-Azetidine hydrochloride

  • MDL number MFCD06804558
  • Cas Number : 871657-49-7
  • Molecular Weight : 151.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/3-1-methylethoxy-azetidine-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9143
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart