ADV465748216 AChemBlock

5-Bromo-7-[trans-4-hydroxycyclohexyl]-2-chloro-7H-pyrrolo[2,3-d]pyrimidine

  • MDL number MFCD27987955
  • Cas Number : 1630906-40-9
  • Molecular Weight : 330.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/5-bromo-7-trans-4-hydroxycyclohexyl-2-chloro-7h-pyrrolo-2-3-d-pyrimidine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9146
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart