ADV465748236 AChemBlock

3-Amino-1-methylpyrazole

  • MDL number MFCD00466340
  • Cas Number : 1904-31-0
  • Molecular Weight : 97.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/3-amino-1-methylpyrazole.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9167
pictograms GHS07
hstatements H313;H303;H333
Availability details for
Quantity
  • Availability
  • Add to Cart