ADV465748247 AChemBlock

4-(Aminomethyl)-2(1H)-pyridinone

  • MDL number MFCD22415181
  • Cas Number : 131052-82-9
  • Molecular Weight : 124.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Room Temp
Physical Form light-yellow solid/white solid
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/4-aminomethyl-2-1h-pyridinone.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3609
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart