ADV465748256 AChemBlock

(R)-4-Boc-2-aminomethylmorpholine hemioxalate

  • MDL number MFCD28166254
  • Cas Number : 1841086-60-9
  • Molecular Weight : 522.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/r-4-boc-2-aminomethylmorpholine-hemioxalate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=B-1049
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart