ADV465748259 AChemBlock

(3S)-3-(4-Hydroxyphenyl)-4-hexynoic acid ethyl ester

  • MDL number MFCD27956896
  • Cas Number : 1292290-97-1
  • Molecular Weight : 232.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/3s-3-4-hydroxyphenyl-4-hexynoic-acid-ethyl-ester.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9190
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart