ADV465748264 AChemBlock

Ethyl 3-isopropyl-1H-pyrazole-5-carboxylate

  • MDL number MFCD03232642
  • Cas Number : 78208-72-7
  • Molecular Weight : 182.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/ethyl-3-isopropyl-1h-pyrazole-5-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9195
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart