ADV465748272 AChemBlock

Benzyl (4-oxocyclopent-2-en-1-yl)carbamate

  • MDL number MFCD28166273
  • Cas Number : 138499-01-1
  • Molecular Weight : 231.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/benzyl-4-oxocyclopent-2-en-1-yl-carbamate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9205
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart