ADV465748273 AChemBlock

(3S,4R)-4-Amino-tetrahydropyran-3-ol hydrochloride

  • MDL number MFCD28166274
  • Cas Number : 1630815-44-9
  • Molecular Weight : 153.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/3s-4r-4-amino-tetrahydropyran-3-ol-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9206
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart