ADV465748275 AChemBlock

8-Bromo-6-chloro-imidazo[1,2-b]pyridazine

  • MDL number MFCD11520890
  • Cas Number : 933190-51-3
  • Molecular Weight : 232.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/8-bromo-6-chloro-imidazo-1-2-b-pyridazine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9208
pictograms GHS07
hstatements H303;H313;H333
Availability details for
Quantity
  • Availability
  • Add to Cart