ADV465748305 AChemBlock

Methyl 3-(methoxycarbonyl)-1-methyl-1H-Pyrrole-2-acetate

  • MDL number MFCD17214380
  • Cas Number : 871819-39-5
  • Molecular Weight : 211.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/methyl-3-methoxycarbonyl-1-methyl-1h-pyrrole-2-acetate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9238
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart