ADV465748307 AChemBlock

1-Boc-3-(2-ethoxy-2-oxoethylidene)pyrrolidine

  • MDL number MFCD28166275
  • Cas Number : 664364-28-7
  • Molecular Weight : 255.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/1-boc-3-2-ethoxy-2-oxoethylidene-pyrrolidine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9240
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart