ADV465748308 AChemBlock

rel-tert-Butyl N-[(1R,4S)-4-hydroxycyclopent-2-en-1-yl]carbamate

  • MDL number MFCD09954747
  • Cas Number : 178152-48-2
  • Molecular Weight : 199.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/rel-tert-butyl-n-1r-4s-4-hydroxycyclopent-2-en-1-yl-carbamate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9241
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart