ADV465748310 AChemBlock

2-Boc-8-oxo-2-azaspiro[4.5]decane

  • MDL number MFCD18711592
  • Cas Number : 1272758-17-4
  • Molecular Weight : 253.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/2-boc-8-oxo-2-azaspiro-4-5-decane.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9243
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart