ADV465748311 AChemBlock

2-Chloro-5-trichloromethylpyridine

  • MDL number MFCD00160145
  • Cas Number : 69045-78-9
  • Molecular Weight : 230.91

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/2-chloro-5-trichloromethylpyridine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9244
pictograms GHS07
hstatements H303;H313;H333
Availability details for
Quantity
  • Availability
  • Add to Cart